Ohaus® Explorer Plus™ EXP Series Precision Toploading Balances
Superb accuracy, seamless set up, intuitive interface, and advanced communication capabilities in one unit to deliver outstanding weighing performance
  • Achieve consistent results with built-in automatic calibration
  • Manage weighing on the intuitive 7” glass touchscreen display
  • Two touchless sensors—set to activate print, zero, tare, or calibrate
  • Motorized leveling system ensures a smooth and straightforward setup
  • Detachable terminal adds flexibility
  • RS-232, USB, and Ethernet connections provide seamless data transfer
  • Supports HID connection to PC or keyboard without drivers
  • Save up to 3000 application sample libraries to easily manage multiple sample profiles
  • Power saving features include auto dim, auto standby, and sleep mode
  • Full metal housing offers durability
Enhance your workflow with the Explorer Plus™ EXP precision toploading balances featuring outstanding weighing performance, intuitive touchscreen display, and advanced communication capabilities. Automatic, user-triggered, and span calibration options ensure precise measurements; the built-in automatic calibration performs self-calibration using an internal mass. A built-in motorized leveling system with a digital bubble automatically levels the balance with the push of a button for a smooth setup.
Easily manage weighing operations on the 7” touchscreen display. An easy-to-clean, chemical-resistant glass panel displays vibrant colors for easy viewing. Detachable terminal offers flexibility—ideal for small labs or placing outside fume hoods. Set terminal status light to indicate overload or underload. Touchless sensors located on the terminal activate often used functions such as print, zero, tare, or calibrate, simply wave your hand over the sensors to activate.
Built-in RS-232, USB (A and C), and Ethernet connections provide seamless data transfer; order an optional dongle for Bluetooth® and Wi-Fi capabilities. Save and recall up to 3000 sample library records to simplify your weighing operation. Each record may contain minimum weight, sample name, sample ID, batch ID, project name, and more. Order an optional fingerprint scanner for added security and a convenient and secure way to log in.
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g)
Availability Pricing
Ohaus EXP223/AD Explorer Plus™ EXP Series Precision Top Loading  Balance, 220g x 1mg, Auto-Draftshield, Touch Screen, 15 Application Modes
New Products
EW-20604-98 Mfr # EXP223/AD
COMPARE
220 0.001
$3,786.58USD / Each
$4,403.00USD / Each
Ohaus EXP423/AD Explorer Plus™ EXP Series Precision Top Loading  Balance, 420g x 1mg, Auto-Draftshield, Touch Screen, 15 Application Modes
New Products
EW-20604-99 Mfr # EXP423/AD
COMPARE
420 0.001
$4,428.14USD / Each
$5,149.00USD / Each
Ohaus EXP623/AD Explorer Plus™ EXP Series Precision Top Loading  Balance, 620g x 1mg, Auto-Draftshield, Touch Screen, 15 Application Modes
New Products
EW-20605-00 Mfr # EXP623/AD
COMPARE
620 0.001
$5,410.26USD / Each
$6,291.00USD / Each
Ohaus EXP1203/AD Explorer Plus™ EXP Series Precision Top Loading  Balance, 1200g x 1mg, Auto-Draftshield, Touch Screen, 15 Application Modes
New Products
EW-20605-01 Mfr # EXP1203/AD
COMPARE
1200 0.001
$6,341.64USD / Each
$7,374.00USD / Each
Ohaus EXP2202 Explorer Plus™ EXP Series Precision Top Loading  Balance, 2200g x 0.01g, Touch Screen, 15 Application Modes
New Products
EW-20605-02 Mfr # EXP2202
COMPARE
2200 0.01
$3,981.80USD / Each
$4,630.00USD / Each
Ohaus EXP4202 Explorer Plus™ EXP Series Precision Top Loading  Balance, 4200g x 0.01g, Touch Screen, 15 Application Modes
New Products
EW-20605-03 Mfr # EXP4202
COMPARE
4200 0.01
$4,576.92USD / Each
$5,322.00USD / Each
Ohaus EXP6202 Explorer Plus™ EXP Series Precision Top Loading  Balance, 6200g x 0.01g, Touch Screen, 15 Application Modes
New Products
EW-20605-04 Mfr # EXP6202
COMPARE
6200 0.01
$5,194.40USD / Each
$6,040.00USD / Each
Ohaus EXP8202 Explorer Plus™ EXP Series Precision Top Loading  Balance, 8200g x 0.01g, Touch Screen, 15 Application Modes
New Products
EW-20605-05 Mfr # EXP8202
COMPARE
8200 0.01
$5,675.14USD / Each
$6,599.00USD / Each
Ohaus EXP12202 Explorer Plus™ EXP Series - Precision Top Loading  Balance, 12000g x 0.01g, Touch Screen, 15 Application Modes
New Products
EW-20605-06 Mfr # EXP12202
COMPARE
12200 0.01
$6,161.04USD / Each
$7,164.00USD / Each
Ohaus EXP6201 Explorer Plus™ EXP Series Precision Top Loading  Balance, 6200g x 0.1g, Touch Screen, 15 Application Modes
New Products
EW-20605-07 Mfr # EXP6201
COMPARE
6200 0.1
$4,319.78USD / Each
$5,023.00USD / Each
Ohaus EXP8201 Explorer Plus™ EXP Series Precision Top Loading  Balance, 8200g x 0.1g, Touch Screen, 15 Application Modes
New Products
EW-20605-08 Mfr # EXP8201
COMPARE
8200 0.1
$4,523.60USD / Each
$5,260.00USD / Each
Ohaus EXP10201 Explorer Plus™ EXP Series Precision Top Loading  Balance, 10200g x 0.1g, Touch Screen, 15 Application Modes
New Products
EW-20605-09 Mfr # EXP10201
COMPARE
10200 0.1
$4,731.72USD / Each
$5,502.00USD / Each
Ohaus EXP24001 Explorer Plus™ EXP Series High-Capacity Top Loading  Balance, 24000g x 0.1g, Touch Screen, 14 Application Modes
New Products
EW-20605-10 Mfr # EXP24001
COMPARE
24000 0.1
$6,038.92USD / Each
$7,022.00USD / Each
Ohaus EXP35001 Explorer Plus™ EXP Series High-Capacity Top Loading  Balance, 35000g x 0.1g, Touch Screen, 14 Application Modes
New Products
EW-20605-11 Mfr # EXP35001
COMPARE
35000 0.1
$7,092.42USD / Each
$8,247.00USD / Each
Ohaus EXP65001 Explorer Plus™ EXP Series High-Capacity Top Loading  Balance, 65000g x 0.1g, Touch Screen, 14 Application Modes
New Products
EW-20605-12 Mfr # EXP65001
COMPARE
65000 0.1
$8,084.86USD / Each
$9,401.00USD / Each
Showing 1- 15 of 15