Ohaus® Frontier™ Multi-Pro Centrifuges
Powerful and versatile Multi-Pro centrifuges handles virtually any lab application
  • High-speed performance at maximum volume
  • Automatic rotor recognition minimize errors
  • Easy-to-turn rotary knob for quick parameter adjustments
  • Large LCD with simple interface
  • Easily switch between rpm and RCF
  • 10 acceleration/deceleration settings
  • 99 memory sets to quickly store and recall commonly used parameters
  • Preset run time from 10 seconds to 99 hours 59 minutes, continuous, or quick spin
  • Motorized lid-locking system prevents the lid from opening during operation
  • Imbalance sensor stops the rotor when imbalance is detected
These multipurpose centrifuges feature high-speed performance, automatic rotor recognition, excellent safety features, and accommodate a broad range of rotors and adapters to provide maximum flexibility. Use this powerful centrifuge for high-speed centrifugation even at maximum volume. The automatic rotor recognition feature conveniently stores all rotor specific data, such as maximum speed and acceleration, saving you time and minimizing errors.
Quickly make parameter adjustments using an easy-to-turn rotary knob. Select from 10 acceleration and 10 deceleration rates to suit your application. Store up to 99 runs with relevant parameters, including the used rotors, to easily repeat the procedures and save time. Safety features include a motorized lid-locking system that prevents the lid from opening during operation and an imbalance sensor that stops the rotor when imbalanced load is detected.
$6,814.00 - $14,694.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Capacity (mL) Max Speed (rpm) Refrigerated
Availability Pricing
Ohaus Frontier FC5718 Multi-Pro Centrifuge, 18,000 rpm, 120V EW-85240-05 Mfr # 30314813
COMPARE
200 18000 No
$6,814.00USD / Each
Ohaus Frontier FC5718R Multi-Pro Refrigerated Centrifuge, 18,000 rpm, 120V EW-85240-06 Mfr # 83042046
COMPARE
200 18000 Yes
$9,916.00USD / Each
Ohaus Frontier FC5916 Multi-Pro Centrifuge, 16,000 rpm, 120V EW-85240-07 Mfr # 30553037
COMPARE
750 16000 No
$10,205.00USD / Each
Ohaus Frontier FC5916R Multi-Pro Refrigerated Centrifuge, 16,000 rpm, 120V EW-85240-08 Mfr # 83042050
COMPARE
750 16000 Yes
$14,404.00USD / Each
Ohaus Frontier FC5718 Multi-Pro Centrifuge, 18,000 rpm, 230V EW-85241-01 Mfr # 30314812
COMPARE
200 18000 No
$6,814.00USD / Each
Ohaus Frontier FC5718R Multi-Pro Refrigerated Centrifuge, 18,000 rpm, 230V EW-85241-02 Mfr # 83042045
COMPARE
200 18000 Yes
$9,916.00USD / Each
Ohaus Frontier FC5720R Multi-Pro High-Speed Refrigerated Centrifuge, 20,000 rpm, 120V EW-85241-48 Mfr # 83042052
COMPARE
200 20000 Yes
$11,303.00USD / Each
Ohaus Frontier FC5720R Multi-Pro High-Speed Refrigerated Centrifuge, 20,000 rpm, 230V EW-85241-49 Mfr # 83042051
COMPARE
200 20000 Yes
$11,303.00USD / Each
Ohaus Frontier FC5830R Multi-Pro High-Speed Refrigerated Centrifuge, 30,000 rpm, 120V EW-85241-50 Mfr # 83042054
COMPARE
250 30000 Yes
$14,694.00USD / Each
Ohaus Frontier FC5830R Multi-Pro High-Speed Refrigerated Centrifuge, 30,000 rpm, 230V EW-85241-51 Mfr # 83042053
COMPARE
250 30000 Yes
$11,303.00USD / Each
Showing 1- 10 of 10