Ohaus Guardian 5000 Digital Stirring Hot Plates
Ensures safe, accurate stirring and heating of your sample
  • Safety system protects from over heating
  • Precise stirring from 50 to 1800 rpm
  • Easy to read LCD display
  • Chemical resistant housing
  • IP32 ingress protection
The Guardian 5000 stirring hot plates are designed to provide powerful heating and mixing capabilities for use in laboratories, pharmacies, schools, businesses, and light industry. Key features combine safety, ease of use, and versatility to ensure reliable performance for everyday applications. Precise stirring from 50 to 1800 rpm is made possible by the design of a powerful motor, strong magnet, and software controlled ramp rate.
Several safety features are built into the hot plates. The SmartHeat™ system provides an upper temperature limit control from 0°C to the rated maximum, preventing overheating. While the early detection system, SafetyHeat™, uses two independent safety controls to continuously monitor the electronics in real-time and automatically shuts off heating before overtemperature conditions occur. Another safety feature is the Run Dry Protection System which shuts off heating and triggers an alarm if the probe-controlled heating is running without the probe tip being submerged in the heating medium. Visual indicators ensure you are aware of potentially dangerous conditions, enhancing overall safety.
The SmartRate™ system allows you to select heating and stirring speeds, from fast and intensive to slow and precise, providing flexibility for various applications. You can adjust the speed or temperature ramping rates for specific needs.
A bright LCD display features large temperature and speed settings, intuitive icons, and clear status indicators, including a prominent hot top indicator, for easy monitoring across the lab. Other features include a RS-232 port for remote control or data logging, green indicator lights when unit is heating and stirring, probe in use icon, single point calibration icon, rotary knob control, two built-in support rod holders, text and symbol HOT caution indicators, large dip angle front panel for easy monitoring from a distance, smooth enclosure surfaces for easy cleaning, audible alarm, IP32 ingress protection, and chemical resistant housing.
$808.00 - $1,321.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Stirring Volume (Liters) Max Temperature (° C) Power (VAC)
Availability Pricing
Ohaus Guardian 5000 e-G52HSRDA Digital Stirring Hot Plate, 5.3" dia, Ceramic Coated Aluminum; 120 VAC EW-85241-07 Mfr # 30541629
COMPARE
20 360 120
$916.00USD / Each
Ohaus Guardian 5000 e-G52HSRDA Digital Stirring Hot Plate, 5.3" dia, Ceramic Coated Aluminum; 230 VAC EW-85241-08 Mfr # 30541630
COMPARE
20 360 230
$916.00USD / Each
Ohaus Guardian 5000 e-G52HS07C Digital Stirring Hot Plate, 7" x 7", Ceramic; 120 VAC EW-85241-09 Mfr # 30541631
COMPARE
20 550 120
$808.00USD / Each
Ohaus Guardian 5000 e-G52HS07C Digital Stirring Hot Plate, 7" x 7", Ceramic; 230 VAC EW-85241-10 Mfr # 30541632
COMPARE
20 550 230
$808.00USD / Each
Ohaus Guardian 5000 e-G52HS10C Digital Stirring Hot Plate, 10" x 10", Ceramic; 120 VAC EW-85241-15 Mfr # 30541637
COMPARE
22 500 120
$1,087.00USD / Each
Ohaus Guardian 5000 e-G52HS10C Digital Stirring Hot Plate, 10" x 10", Ceramic; 230 VAC EW-85241-16 Mfr # 30541638
COMPARE
22 500 230
$1,087.00USD / Each
Ohaus Guardian 5000 e-G52HS07C Digital Stirring Hot Plate Kit, 7" x 7", Ceramic; 120 VAC EW-85241-17 Mfr # 30981778
COMPARE
20 550 120
$1,075.00USD / Each
Ohaus Guardian 5000 e-G52HS07C Digital Stirring Hot Plate Kit, 7" x 7", Ceramic; 230 VAC EW-85241-18 Mfr # 30981779
COMPARE
20 550 230
$1,075.00USD / Each
Ohaus Guardian 5000 e-G52HS10C Digital Stirring Hot Plate Kit, 10" x 10", Ceramic; 120 VAC EW-85241-21 Mfr # 30981782
COMPARE
22 500 120
$1,321.00USD / Each
Ohaus Guardian 5000 e-G52HS10C Digital Stirring Hot Plate Kit, 10" x 10", Ceramic; 230 VAC EW-85241-22 Mfr # 30981783
COMPARE
22 500 230
$1,321.00USD / Each
Ohaus Guardian 5000 e-G52HSRDA Digital Stirring Hot Plate Kit, 5.3" dia, Ceramic Coated Aluminum; 120 VAC EW-85241-23 Mfr # 30981776
COMPARE
22 360 120
$1,166.00USD / Each
Showing 1- 11 of 11