Ohaus Heavy Duty Digital Vortex Mixers
Increase productivity with continuous-duty operation
  • Safe to mix at high speeds
  • Variable speeds up to 3500 rpm
  • Effective speed control from low to high-speed mixing
  • Compact design takes up less benchtop space
  • 2 year warranty
The heavy-duty vortex mixers are engineered for durability and efficiency, featuring a high-quality German motor designed for continuous operation. A unique locking design ensures safe operation across the entire speed range.
Capable of handling loads up to 1.1 kg, the vortex mixers are ideal for demanding tasks. A versatile speed control allows you to adjust from gentle shaking to vigorous vertexing, accommodating a wide range of sample types.
These mixers are perfect for laboratories needing reliable performance and flexibility in their mixing applications. The digital model provides precise speed control and a timer for consistent results, while non-digital model offers a cost-effective alternative with variable speed settings.
$1,274.00 - $1,482.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Max Load (lbs) Max Speed (rpm)
Availability Pricing
Ohaus Heavy Duty Digital Vortex Mixer, 300 to 3500 rpm; 120 VAC EW-53027-60 Mfr # VXHDDG
COMPARE
Analog 2.5 3500
$1,482.00USD / Each
Ohaus Heavy Duty Analog Vortex Mixer, 300 to 3500 rpm; 120 VAC EW-53027-61 Mfr # VXHDAL
COMPARE
Digital 2.5 3500
$1,274.00USD / Each