Ohaus PR Series Toploading Balances
Reliable performance at an affordable price
  • Bright, easy-to-read backlit LCD
  • RS-232 interface allows connection to printer or PC
  • Small footprint saves desktop space
  • Selectable environmental filters compensate for harsh environments
  • Removable stainless-steel pan for easy cleaning
These toploading balances provide basic weighing, parts counting, and percent weighing applications. The backlit LCD and easy-to-use interface make these balances simple to operate. A standard RS-232 port provides communication with point-of-sale systems or printers.
The balances are designed with durable ABS housing and a removable stainless-steel pan. Additional features include integrated weigh-below hook, security bracket, calibration lock, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, and stability indicator.
Designed for accuracy and repeatability in weighting applications, these balances are ideal for laboratory, industrial, and educational settings at an economical price.
ITEM(S) ON SALE
$921.06 - $1,410.40USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Repeatability (g)
Availability Pricing
Ohaus PR523/E Precision Toploading Balance 520g  x 0.001g  External Calibration EW-11601-22 Mfr # PR523/E
COMPARE
520 0.001 0.001
$1,339.02USD / Each
$1,557.00USD / Each
Ohaus PR2202/E Precision Toploading Balance 2200g  x 0.01g  External Calibration EW-11601-23 Mfr # PR2202/E
COMPARE
2200 0.001 0.01
$1,179.92USD / Each
$1,372.00USD / Each
Ohaus PR4202/E Precision Toploading Balance 4200g  x 0.01g  External Calibration EW-11601-24 Mfr # PR4202/E
COMPARE
4200 0.001 0.01
$1,410.40USD / Each
$1,640.00USD / Each
Ohaus PR4201/E Precision Toploading Balance 4200g  x 0.1g  External Calibration EW-11601-25 Mfr # PR4201/E
COMPARE
4200 0.1 0.1
$1,064.68USD / Each
$1,238.00USD / Each
Ohaus PR1602/E Precision Toploading Balance 1600g  x 0.01g  External Calibration EW-11601-26 Mfr # PR1602/E
COMPARE
1600 0.001 0.01
$1,035.44USD / Each
$1,204.00USD / Each
Ohaus PR2201/E Precision Toploading Balance 2200g  x 0.1g  External Calibration EW-11601-27 Mfr # PR2201/E
COMPARE
2200 0.1 0.1
$921.06USD / Each
$1,071.00USD / Each
Ohaus PR223/E Precision Toploading Balance 220g  x 0.001g  External Calibration EW-11601-28 Mfr # PR223/E
COMPARE
220 0.001 0.001
$1,122.30USD / Each
$1,305.00USD / Each
Showing 1- 7 of 7