Ohaus® Standard-Duty Incubating Orbital Shakers
Incubator and shaker in one unit saves valuable benchtop space
  • Control and view speed, temperature, and time independently
  • 3 mm (0.1”) orbital motion
  • Triple eccentric drive systems provide stable orbital motion
  • Microprocessor-control offers accurate shaking speed
  • View actual/setpoint temperatures
  • Count up/count down timer
These standard-duty incubating orbital shakers are perfect for immunoassay, hybridization, PCR, cell cultures, and enzyme reaction applications. Independent controls for speed, temperature, and time and an intuitive touchpad make it easy to set and view the parameters. Digital speed control offers reliable and repeatable shaking speed. Heat samples up to 65 °C; a dome cover helps maintain uniform chamber temperature. Built with operator safety in mind, a “Caution Hot” indicator illuminates if the chamber air temperature is above 40 °C (104 °F). Set the timer to display accumulated time (continuous mode) or remaining time (timed mode) from 1 second to 9999 minutes. Use the included aluminum tray to hold clamps, trays, tacks, or plates for secure shaking.
$3,472.00 - $3,852.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Operating Temperature (° C) Max Speed (rpm) Description
Availability Pricing
Ohaus Standard-Duty Incubating Orbital Shaker, 3 mm Orbit EW-53027-82 Mfr # ISLD04HDG
COMPARE
65 1200 Standard-Duty Incubating Orbital Shaker, 3 mm Orbit
$3,852.00USD / Each
Ohaus Standard-Duty Incubating Orbital Microplate Shaker, 3 mm Orbit EW-53027-83 Mfr # ISLDMPHDG
COMPARE
65 1200 Standard-Duty Incubating Orbital Microplate Shaker, 3 mm Orbit
$3,472.00USD / Each
Ohaus Standard-Duty Incubating Orbital Microplate Shaker with Opaque Lid, 3 mm Orbit EW-53027-84 Mfr # ISLDMPHDGL
COMPARE
65 1200 Standard-Duty Incubating Orbital Microplate Shaker with Opaque Lid, 3 mm Orbit
$3,472.00USD / Each