Omni International Bead Ruptor 12 Bead Mill Homogenizer and Bead Kits
Disposable tubes prevent cross-contamination
  • Homogenize up to twelve 2-mL or 0.5-mL tubes at one time
  • Quick processing inhibits sample degradation
The Omni Bead Ruptor 12 homogenizer simultaneously processes samples, from 25 µL to 1.8 mL. Unit provides efficient and consistent homogenization, as well as high yield, by vigorously and uniformly shaking the tubes. A small footprint optimizes bench space in crowded labs. Convenient front-loading tube holder position permits quick and easy loading of samples. No cool-down between runs means nonstop processing and minimal down time.
Five different bead types allow processing of different samples. Ceramic beads are ideal for soft animal, plant or human tissues. Metal beads are ideal for hard tissue types. Glass beads are ideal for processing microorganisms. Garnet beads are ideal for soil. Carbide beads are ideal for microbial and soil RNA processing.
$9,295.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Max Speed (rpm) Min Sample Size (mL)
Availability Pricing
Omni International 19-050 Bead Ruptor 12 Bead Mill Homogenizer; 110-230 VAC, 50/60 Hz EW-04728-01 Mfr # 19-050
COMPARE
1.8
$9,295.00USD / Each