Pacer DA400 Series Precision Vane Anemometers with 2.75” Vane
Highly accurate even down to ultralow flow rates
  • Rugged metal rotating vane is ideal for industrial use
  • Includes NIST-traceable calibration certificate
Pacer precision vane anemometers are simple to use yet offer excellent air velocity accuracy even at low flow rates. Industrial grade enclosure and probe lets you measure airflow in extreme environments from –20 to 100°C (–4 to 212°F). All models feature Min/Max (air and temp only), data Hold, and auto power off. Large display with backlight lets you view readings easily.
$1,265.00 - $1,640.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Air Volume (CFM) Max Temperature (° C) RH Accuracy
Availability Pricing
Pacer DA430 Precision Vane Thermoanemometer with 2.75" Vane and Temperature/Humidity Probe EW-10001-15 Mfr # 10202
COMPARE
80 ±2.0% RH
$1,290.00USD / Each
Pacer DA400 Precision Vane Anemometer with USB Output EW-10001-20 Mfr # 10199+10216
COMPARE
$1,265.00USD / Each
Pacer DA410 Precision Vane Anemometer with 2.75" Vane, Volume Calculation, and USB Output EW-10001-25 Mfr # 10200+10216
COMPARE
$1,350.00USD / Each
Pacer DA420 Precision Vane Thermoanemometer with 2.75" Vane and USB Output EW-10001-30 Mfr # 10201+10223
COMPARE
100
$1,420.00USD / Each
Pacer DA430 Precision Vane Thermoanemometer with 2.75" Vane, Temperature/Humidity Probe, and USB Output; IP65 EW-10001-40 Mfr # 10202+10223+10227
COMPARE
80 ±2.0% RH
$1,640.00USD / Each