PendoTECH® Single-Use Pressure Sensors™
Accurate, reliable, and cost-effective pressure monitoring for bioprocessing
  • Wide compatibility with hose-barb, sanitary flange, and luer fittings
  • Highly accurate across a broad pressure range (-11.5 to 75 psi)
  • Unobstructed fluid path minimizes hold-up volume and dead legs
  • Pre-tested for critical quality attributes and USP Class VI compliant
  • Compatible with gamma and x-ray irradiation for sterilization
PendoTECH® Single-Use Pressure Sensors™ provide precise monitoring of gas and liquid pressure in critical bioprocessing applications such as filtration, chromatography, and bioreactor operations. Designed with MEMS-HAP™ high-accuracy sensor chips, they deliver reliable, real-time pressure readings while eliminating contamination risks. Available in multiple connection types, these sensors feature a seamless fluid path and are 100% quality tested to ensure peak performance. They integrate easily with PendoTECH PressureMAT® monitors, offering a cost-effective, single-use alternative for pressure measurement in regulated environments.
$81.90 - $157.50USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Process Connection Wetted Materials
Availability Pricing
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, Luer Inlet/Outlet
Always in Stock
EW-59900-11 Mfr # 30961415
COMPARE
Luer Polysulfone
$81.90USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 1/8" Hose Barb EW-59900-12 Mfr # PREPS-N-012
COMPARE
1/8" hose barb Polysulfone
$94.50USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 1/4" Hose Barb
Always in Stock
EW-59900-13 Mfr # PREPS-N-025
COMPARE
1/4" hose barb Polysulfone
$102.00USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 3/8" Hose Barb EW-59900-14 Mfr # PREPS-N-038
COMPARE
3/8" hose barb Polysulfone
$110.25USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 1/2" Hose Barb EW-59900-15 Mfr # PREPS-N-050
COMPARE
1/2" hose barb Polysulfone
$105.00USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone,  3/4" Hose Barb EW-59900-16 Mfr # PREPS-N-075
COMPARE
3/4" hose barb Polysulfone
$115.50USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone,  1" Sanitary Flange Inlet/Outlet EW-59900-17 Mfr # PREPS-N-1-1
COMPARE
1" sanitary flange Polysulfone
$157.50USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone,  1/2" Sanitary Flange Inlet/Outlet EW-59900-18 Mfr # PREPS-N-5-5
COMPARE
1/2" sanitary flange Polysulfone
$136.50USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polycarbonate, 1/4" Hose Barb EW-59900-19 Mfr # PRESS-N-025
COMPARE
1/4" hose barb Polycarbonate
$88.20USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polycarbonate,  3/8" Hose Barb EW-59900-20 Mfr # PRESS-N-038
COMPARE
3/8" hose barb Polycarbonate
$91.40USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polycarbonate, 1/2" Hose Barb EW-59900-21 Mfr # PRESS-N-050
COMPARE
1/2" hose barb Polycarbonate
$91.40USD / Each
PendoTech Single-Use Pressure Sensor, Sterile, Polycarbonate, Luer Inlet/Outlet EW-59900-27 Mfr # 30961417
COMPARE
Luer Polycarbonate
$89.30USD / Each
Showing 1- 12 of 12