Phase II PHT-1800 Portable Digital Hardness Tester
The most accurate, economically priced portable hardness tester on the market
  • Auto identification of Impact Device
  • Automatic conversions to Brinell, Rockwell B & C, Vickers, and Shore
  • Automatic mean value as well as Min/Max values
  • Memory capacity of up to 100 groups
  • Large LCD display with back light
  • USB ouput
  • Low battery indicator
  • Easy-to-use keypad operation
This portable digital hardness tester is designed to accurately measure the hardness of metal materials and includes features normally only found on high-priced full-function models. With its fast test speeds, precision accuracy, and memory capability and output, this hardness tester is perfect for applications in the QC lab or the out in the field.
$2,092.65USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Display Power Range
Availability Pricing
Phase-II PHT-1800 Portable Digital Hardness Tester 54103-01 EW-54103-01 Mfr # PHT-1800
COMPARE
Large LCD display with back light Four AA batteries (included) 200 to 960 HL, 20 to 70 HRC, 20 to 99 HRB, 40 to 650 HB, 80 to 970 HV, 30 to 100 HS
$2,092.65USD / Each