PYREX Brand 5340 Filtering Flasks
These heavy wall flasks with tabulation are blown in special molds in shapes designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers. Tubulation OD on flasks up to 1000 mL is 10 mm. Tubulation OD for 2000-mL and 4000-mL flasks is 12.5 mm.
$210.00 - $1,205.00USD / Pkg of 6
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Stopper Number Accepted
Availability Pricing
Pyrex 5340-250 5340 Filtering Flasks, 250 mL; 6/Pk
Buy More Save More
EW-34557-01 Mfr # 5340-250
COMPARE
250 25 6
$236.00USD / Pkg of 6
Pyrex 5340-500 5340 Filtering Flasks, 500 mL; 6/Pk
Buy More Save More
EW-34557-02 Mfr # 5340-500
COMPARE
500 50 7
$338.00USD / Pkg of 6
Pyrex 5340-1L 5340 Filtering Flasks, 1000 mL; 6/Pk
Buy More Save More
EW-34557-03 Mfr # 5340-1L
COMPARE
1000 50 8
$618.00USD / Pkg of 6
Pyrex 5340-2L 5340 Filtering Flasks, 2000 mL; 1/Pk
Buy More Save More
EW-34557-04 Mfr # 5340-2L
COMPARE
2000 200 9
$210.00USD / Pkg of 1
Pyrex 5340-4L 5340 Filtering Flasks, 4000 mL; 1/Pk
Buy More Save More
EW-34557-05 Mfr # 5340-4L
COMPARE
4000 500 12
$305.00USD / Pkg of 1
Pyrex 5340-1L 5340 Filtering Flasks, 1000 mL; 12/Cs
Buy More Save More
EW-34557-23 Mfr # 5340-1L
COMPARE
1000 50 8
$849.00USD / Case of 12
Pyrex 5340-4L 5340 Filtering Flasks, 4000 mL; 6/Cs
Buy More Save More
EW-34557-25 Mfr # 5340-4L
COMPARE
4000 500 12
$1,205.00USD / Case of 6
Showing 1- 7 of 7