Quick Mist Dispenser Bottle, HDPE
Gives an excellent, precise mist with large pattern!
  • Chemically resistant polypropylene trigger
  • Easy to clean valve and intake
Trigger sprayer is manufactured with a chemically resistant polypropylene and includes a plastic strainer. Reusable sprayer features easy to clean valve and intake. Ideal for thin liquids. Not suitable for use with food products.
$35.10 - $43.10USD / Pkg of 4
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Capacity (mL) Overall Height (in)
Availability Pricing
Quick Mist HDPE Dispenser/Spray Bottle, 16 oz (500 mL), 4/Pk
Buy More Save More
EW-06091-01 Mfr # 605144
COMPARE
No 500 7 7/8
$35.10USD / Pkg of 4
Quick Mist HDPE Dispenser/Spray Bottle, 32 oz (1 L), 4/Pk
Buy More Save More
EW-06091-03 Mfr # 605154
COMPARE
No 1000 9 3/8
$43.10USD / Pkg of 4