Readycult Coliforms Test in Finished Water
EPA approved rapid presence/absence test for the simultaneous detection of total coliforms and E. coli in drinking water
  • EPA-approved testing protocol
  • Rapid, one-step method with a 24-hour incubation response time for total coliform presence
  • A quick second step for confirmation of the presence of E.coli
Save both time and expense by avoiding the traditional coliform & E. coli testing methods. Readycult® tests allow you to respond more quickly to potentially dangerous situations when coliforms and E. coli are found to be present in water samples. As an added bonus, you'll never have to worry about product integrity because the Readycult® resealable packs do not require refrigeration and have a shelf life of three years (post manufacturing date).
Application is extremely simple, just open a Readycult Just add a Readycult snap pack into your sample and mix, then incubate for 24 hours at 35-37oC. If there is no color change, the test is negative. Any color change to blue-green confirms the presence of total coliforms. Presence of E. Coli creates a blue fluorescence (MUG reaction) under 365nm UV-light. Indole test 36152-18 can be directly performed in the same broth for E. Coli confirmation.
$167.75USD / Pkg of 20
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Tests Included Test Type
Availability Pricing
Readycult 1.01298.0001 Coliforms Test, snap pack, 20/pk EW-36152-10 Mfr # 1.01298.0001
COMPARE
20 Coliforms, E. coli
$167.75USD / Pkg of 20