Reagents® Conductivity Standard 4000 µS/cm
Manufactured using the highest quality raw materials in ISO 9001 facility
  • 4,000µS/cm, (Range 3,990 to 4,010µS/cm)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$61.40 - $194.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Conductivity Standard 4000 µS/cm, 1L EW-78915-23 Mfr # CC940000-1A
COMPARE
Conductivity 1000 Conductivity Standard 4000 µS/cm, 1L,
$61.40USD / Each
Reagents Conductivity Standard 4000 µS/cm, 4L EW-78920-95 Mfr # CC940000-4A
COMPARE
Conductivity 4000 Conductivity Standard 4000 µS/cm, 4L,
$194.00USD / Each