Reagents® EDTA Titrant, 0.01M, Standardized

Manufactured using the highest quality raw materials in ISO 9001 facility
  • M/100, 0.02N, N/50, Assay 0.00995 to 0.01005 Molar, Aqueous
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$47.50 - $413.50USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents EDTA Titrant, 0.01M, 1L, Standardized EW-78915-36 Mfr # CE250400-1A
COMPARE
Standardized 1000 EDTA Titrant, 0.01M, 1L, Standardized
$47.50USD / Each
Reagents EDTA Titrant, 0.01M, 4L, Standardized, 1.02 Density EW-78921-07 Mfr # CE250400-4A
COMPARE
Standardized 4000 EDTA Titrant, 0.01M, 4L, Standardized
$113.25USD / Each
Reagents EDTA Titrant, 0.01M, 10L, Standardized EW-78925-72 Mfr # CE250400-10F
COMPARE
Standardized 10000 EDTA Titrant, 0.01M, 10L, Standardized
$268.25USD / Each
Reagents EDTA Titrant, 0.01M, 20L, Standardized EW-78927-26 Mfr # CE250400-20F
COMPARE
Standardized 20000 EDTA Titrant, 0.01M, 20L, Standardized
$413.50USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat