Reagents® EDTA Titrant, 0.25M, Standardized
Manufactured using the highest quality raw materials in ISO 9001 facility
  • M/25, N/2, Assay 0.2495 to 0.2505 Molar, Aqueous
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$115.00 - $151.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents EDTA Titrant, 0.25M, 500mL, Standardized EW-78911-17 Mfr # CE251000-500A
COMPARE
Standardized 500 EDTA Titrant, 0.25M, 500mL, Standardized
$115.00USD / Each
Reagents EDTA Titrant, 0.25M, 1L, Standardized EW-78915-40 Mfr # CE251000-1A
COMPARE
Standardized 1000 EDTA Titrant, 0.25M, 1L, Standardized
$151.25USD / Each