Reagents® Formaldehyde, 37%, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$44.70 - $405.50USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Formaldehyde, 37%, 500mL, Purified EW-78911-45 Mfr # C2208500-500A
COMPARE
Formaldehyde 500 Formaldehyde, 37%, 500mL, Purified
$44.70USD / Each
Reagents Formaldehyde, 37%, 1L, Purified EW-78915-53 Mfr # C2208500-1A
COMPARE
Formaldehyde 1000 Formaldehyde, 37%, 1L, Purified
$73.90USD / Each
Reagents Formaldehyde, 37%, 4L, Purified EW-78921-21 Mfr # C2208500-4A
COMPARE
Formaldehyde 4000 Formaldehyde, 37%, 4L, Purified
$121.75USD / Each
Reagents Formaldehyde, 37%, 20L, Purified EW-78928-81 Mfr # C2208500-20F
COMPARE
Formaldehyde 20000 Formaldehyde, 37%, 20L, Purified
$405.50USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat