Reagents® Hydrochloric Acid, 0.010N, Standardized
Manufactured using the highest quality raw materials in ISO 9001 facility
  • N/100, Assay 0.00995 to 0.01005 Normal, Aqueous
Hydrochloric Acid, 1L, 0.01N ConcentratIon, Colorless, Liquid, 100°C Boiling Point, 0°C Melting Point, 2 pH Range, 1 Density, 24 Months Shelf Life.
$39.20 - $82.30USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Hydrochloric Acid, 0.010N, 1L, Standardized EW-78915-59 Mfr # C7101010-1A
COMPARE
Standardized 1000 Hydrochloric Acid, 0.010N, 1L, Standardized
$39.20USD / Each
Reagents Hydrochloric Acid, 0.010N, 4L, Standardized EW-78921-27 Mfr # C7101010-4A
COMPARE
Standardized 4000 Hydrochloric Acid, 0.010N, 4L, Standardized
$82.30USD / Each