Reagents® Phosphate Buffer, Standardized, pH 7.2

Manufactured using the highest quality raw materials in ISO 9001 facility
  • pH 7.2
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$64.40 - $219.25USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Phosphate Buffer, 500mL, Standardized EW-78912-72 Mfr # C7271150-500A
COMPARE
Buffer 500 Phosphate Buffer, 500mL, Standardized
$64.40USD / Each
Reagents Phosphate Buffer, 1L, Standardized EW-78916-24 Mfr # C7271150-1A
COMPARE
Buffer 1000 Phosphate Buffer, 1L, Standardized
$97.10USD / Each
Reagents Phosphate Buffer, 4L, Standardized EW-78923-29 Mfr # C7271150-4A
COMPARE
Buffer 4000 Phosphate Buffer, 4L, Standardized
$219.25USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat