Reagents® Sodium Bisulfate Anhydrous

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless/white solid
  • Density: 2.74
  • Solubility information: 285 g/L at 25 °C (77 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$36.50 - $114.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Sodium Bisulfate Anhydrous, Wide-Mouth Amber Glass Bottle, 100 g EW-62285-61 Mfr # C2309500-100C2
COMPARE
Salts Wide-mouth amber glass bottle 100
$36.50USD / Each
Reagents Sodium Bisulfate Anhydrous, Wide-Mouth Container, 500 g EW-78913-59 Mfr # C2309500-500A5
COMPARE
Salts Wide-mouth container 500
$114.50USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat