Reagents® Sodium Hydroxide, 0.10N, Standardized

Manufactured using the highest quality raw materials in ISO 9001 facility
  • N/10
  • Assay 0.0995 to 0.1005 Normal
  • Aqueous
  • Colorless liquid
  • Density: 1
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$18.80 - $1,065.00USD / Each

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Sodium Hydroxide, 0.10N, Standardized, Natural Poly Bottle, 100 mL EW-62288-28 Mfr # C7318000-100A
COMPARE
Standardized Poly bottle 100
$18.80USD / Each
Reagents Sodium Hydroxide, 0.10N, Standardized, Drum, 200 L EW-62288-29 Mfr # C7318000-200E
COMPARE
Standardized Drum 200000
$1,065.00USD / Each
Reagents Sodium Hydroxide, 0.10N, Standardized, Natural Poly Bottle, 500 mL EW-78913-83 Mfr # C7318000-500A
COMPARE
Standardized Poly bottle 500
$27.70USD / Each
Reagents Sodium Hydroxide, 0.10N, Standardized, Natural Poly Bottle, 1 L EW-78916-82 Mfr # C7318000-1A
COMPARE
Standardized Poly bottle 1000
$43.40USD / Each
Reagents Sodium Hydroxide, 0.10N, Standardized, Natural Poly Bottle, 4 L EW-78921-98 Mfr # C7318000-4A
COMPARE
Standardized Poly bottle 4000
$84.60USD / Each
Reagents Sodium Hydroxide, 0.10N, Standardized, Cubitainer® Package, 4 L EW-78921-99 Mfr # C7318000-4F
COMPARE
Standardized Cubitainer® package 4000
$88.80USD / Each
Reagents Sodium Hydroxide, 0.10N, Standardized, Cubitainer® Package, 10 L EW-78926-03 Mfr # C7318000-10F
COMPARE
Standardized Cubitainer® package 10000
$165.75USD / Each
Reagents Sodium Hydroxide, 0.10N, Standardized, Cubitainer® Package, 20 L EW-78927-60 Mfr # C7318000-20F
COMPARE
Standardized Cubitainer® package 20000
$244.50USD / Each
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat