Reagents® Sodium Thiosulfate, 0.010N, Standardized
Manufactured using the highest quality raw materials in ISO 9001 facility
  • N/100, Assay 0.0099 to 0.0101 Normal, Aqueous
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$47.00 - $233.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Sodium Thiosulfate, 0.010N, 1L, Standardized EW-78918-39 Mfr # CS000100-1C
COMPARE
Standard 1000 Sodium Thiosulfate, 0.010N, 1L, Standardized
$47.00USD / Each
Reagents Sodium Thiosulfate, 0.010N, 4L, Standardized EW-78924-46 Mfr # CS000100-4C
COMPARE
Standard 4000 Sodium Thiosulfate, 0.010N, 4L, Standardized
$124.00USD / Each
Reagents Sodium Thiosulfate, 0.010N, 10L, Standardized EW-78926-10 Mfr # CS000100-10F
COMPARE
Standard 10000 Sodium Thiosulfate, 0.010N, 10L, Standardized
$233.50USD / Each