ResinTech AF Pro Series Drop-in Filters
Eliminate harmful contaminants and improve drinking water
  • Fits standard POE and POU housings
  • For residential & industrial applications
  • Oversized cartridge for maximum media fill
  • Lot numbered for component traceability
  • Made in the USA!
These AF-series filter cartridges are designed to fit standard 10- or 20-inch point-of-use and point-of-entry housings, utilizing a wide variety of medias that eliminate harmful contaminants and improve drinking water. Housings are injection molded and made of high-grade materials.
Cartridges are certified with the WQA Gold Seal, ensuring you receive the highest quality products that meet the strictest manufacturing standards. The WQA Gold Seal program assures that cartridge materials and construction are tested and certified to meet NSF/ANSI certification standards.
Choose from a wide range of filter cartridge options depending on the specific contaminants you need to eliminate.
$20.90 - $310.55USD / Each
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Compatible with Max Flow Rate (GPH)
Availability Pricing
ResinTech AF-10-1052 Coconut Shell Carbon Filter Cartridge (10" Slimline) EW-99284-00 Mfr # AF-10-1052
COMPARE
4000 10" Standard Housing 60
$24.72USD / Box of 1
ResinTech AF-20-1052 Coconut Shell Carbon Filter Cartridge (20" Slimline) EW-99284-01 Mfr # AF-20-1052
COMPARE
9000 20" Standard Housing 120
$38.30USD / Box of 1
ResinTech AF-10-1046-BB Coconut Shell Carbon Filter Cartridge (10" Big Blue) EW-99284-02 Mfr # AF-10-1046-BB
COMPARE
13000 10" Standard Housing 240
$60.00USD / Box of 1
ResinTech AF-10-1053 Chloramine Reduction Filter Cartridge (10" Slimline) EW-99284-08 Mfr # AF-10-1053
COMPARE
2000 10" Standard Housing 60
$49.95USD / Box of 1
ResinTech AF-10-3326-BB Scale Prevention Filter Cartridge (10" Big Blue) EW-99284-17 Mfr # AF-10-3326-BB
COMPARE
10" Standard Housing 180
$95.00USD / Box of 1
ResinTech AF-20-3327-BB Scale Prevention Filter Cartridge (20" Big Blue) EW-99284-18 Mfr # AF-20-3327-BB
COMPARE
20" Standard Housing 300
$147.20USD / Box of 1
ResinTech AF-10-3232 pH Neutralization Filter Cartridge (10" Slimline) EW-99284-19 Mfr # AF-10-3232
COMPARE
7500 10" Standard Housing 30
$20.90USD / Box of 1
ResinTech AF-10-3610 Nitrate Reduction Filter Cartridge (10" Slimline) EW-99284-23 Mfr # AF-10-3610
COMPARE
475 10" Standard Housing 6
$69.40USD / Box of 1
ResinTech AF-10-3610-BB Nitrate Reduction Filter Cartridge (10" Big Blue) EW-99284-25 Mfr # AF-10-3610-BB
COMPARE
1200 10" Standard Housing 15
$162.55USD / Box of 1
ResinTech AF-20-3610-BB Nitrate Reduction Filter Cartridge (20" Big Blue) EW-99284-26 Mfr # AF-20-3610-BB
COMPARE
2500 20" Standard Housing 30
$310.55USD / Box of 1
ResinTech AF-10-4030 High Purity Color-Changing DI Filter Cartridge (10" Slimline) EW-99284-49 Mfr # AF-10-4030
COMPARE
1320 10" Standard Housing 18
$60.30USD / Each
ResinTech AF-10-4040 High Capacity Color-Changing DI Filter Cartridge (10" Slimline) EW-99284-51 Mfr # AF-10-4040
COMPARE
1600 10" Standard Housing 18
$60.30USD / Each
ResinTech AF-20-4040 High Capacity Color-Changing DI Filter Cartridge (20" Slimline) EW-99284-52 Mfr # AF-20-4040
COMPARE
3200 20" Standard Housing 18
$120.55USD / Each
ResinTech AF-10-4010 Semiconductor Grade DI Filter Cartridge (10" Slimline) EW-99284-53 Mfr # AF-10-4010
COMPARE
1320 10" Standard Housing 18
$53.85USD / Each
ResinTech AF-20-4010 Semiconductor Grade DI Filter Cartridge (20" Slimline) EW-99284-54 Mfr # AF-20-4010
COMPARE
2640 20" Standard Housing 36
$102.40USD / Each
ResinTech AF-10-4010-BB Semiconductor Grade DI Filter Cartridge (10" Big Blue) EW-99284-55 Mfr # AF-10-4010-BB
COMPARE
3000 10" Standard Housing 48
$115.60USD / Each
ResinTech AF-20-4010-BB Semiconductor Grade DI Filter Cartridge (20" Big Blue) EW-99284-56 Mfr # AF-20-4010-BB
COMPARE
6800 20" Standard Housing 90
$221.05USD / Each
ResinTech AF-10-4011 Premium Purity LTOC DI Filter Cartridge (10" Slimline) EW-99284-57 Mfr # AF-10-4011
COMPARE
1320 10" Standard Housing 18
$65.90USD / Each
ResinTech AF-20-4011-BB Ultra High-Purity LTOC DI Filter Cartridge (20" Big Blue) EW-99284-60 Mfr # AF-20-4011-BB
COMPARE
6800 20" Standard Housing 90
$271.95USD / Each
ResinTech AF-20-4020 Ultra High-Purity & Low Odor DI Filter Cartridge (20" Slimline) EW-99284-62 Mfr # AF-20-4020
COMPARE
2640 20" Standard Housing 36
$116.60USD / Each
Showing 1 - 20 of 29