Saint-Gobain Pure-Fit® SIB® 3/4" Mini to Hose Barb Fittings, Straight Adapter, Polypropylene
Smooth inner bore ensures seamless fluid transition between tubing and fittings
  • Unique internal design eliminates dead space
  • No areas of entrapment—prohibiting a potential for bacterial growth
  • Use with Pure-Fit and BarbLock® retainers for leak-free connections
  • Easily transition from sanitary fitting to a barb connection
  • Steriliza by gamma irradiation
  • Animal-derived component free and BPA free
  • Manufactured and packaged in Class 8 cleanroom
  • USP <88> Class VI and USP <87> compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Eliminate dead space and entrapment, a potential breeding ground for bacterial growth, with smooth inner bore (SIB) fittings. SIB fittings provide a seamless, “zero” transition between tubing and fittings, maintaining complete fluid integrity. Straight adapters allow easy transition from a sanitary fitting system to a barbed connection. All fittings feature a 3/4" mini fitting and a barbed fitting—order a gasket and clamp to complete your sanitary connection. Use with Pure-Fit or BarbLock retainer assembly that provides 360° compression to ensure a leak-free connection.
$282.50 - $325.00USD / Pkg of 25
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Sanitary Clamp Size (Port 1) (InchShort)
Availability Pricing
Saint Gobain Pure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 1/8"; 25/Pk EW-50143-12 Mfr # PFMS125PPAF
COMPARE
1/8 3/4
$282.50USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 1/4"; 25/Pk EW-50143-26 Mfr # PFMS250PPAF
COMPARE
1/4 3/4
$293.75USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 5/16"; 25/Pk EW-50143-32 Mfr # PFMS312PPAF
COMPARE
5/16 3/4
$303.00USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 3/8"; 25/Pk EW-50143-38 Mfr # PFMS375PPAF
COMPARE
3/8 3/4
$321.50USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 1/2"; 25/Pk EW-50143-46 Mfr # PFMS500PPAF
COMPARE
1/2 3/4
$325.00USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Sanitary to Hose Barb Fitting, Straight Adapter, Polypropylene, 3/4" Mini x 5/8"; 25/Pk EW-50143-49 Mfr # PFMS625PPAF
COMPARE
5/8 3/4
$310.50USD / Pkg of 25