Saint-Gobain Pure-Fit® SIB® Hose Barb Fittings, Cross Union, Polypropylene
Smooth inner bore ensures seamless fluid transition between tubing and fittings
  • Unique internal design eliminates dead space
  • No areas of entrapment—prohibiting a potential for bacterial growth
  • Use with Pure-Fit and BarbLock® retainers for leak-free connections
  • Easily combine or split fluid flow with a cross union
  • Steriliza by gamma irradiation
  • Animal-derived component free and BPA free
  • Manufactured and packaged in Class 8 cleanroom
  • USP <88> Class VI and USP <87> compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Eliminate dead space and entrapment, a potential breeding ground for bacterial growth, with smooth inner bore (SIB) fittings. SIB fittings provide a seamless, “zero” transition between tubing and fittings, maintaining complete fluid integrity. Use with Pure-Fit or BarbLock retainer assembly that provides 360° compression to ensure a leak-free connection.
$266.75 - $307.50USD / Pkg of 25
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in)
Availability Pricing
Saint Gobain Pure-Fit® SIB® Hobe Barb Fitting, Cross Union, Polypropylene, 1/8"; 25/Pk EW-50143-15 Mfr # PFX125PPAF
COMPARE
1/8
$266.75USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hobe Barb Fitting, Cross Union, Polypropylene, 3/16"; 25/Pk EW-50143-18 Mfr # PFX187PPAF
COMPARE
3/16
$274.25USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hobe Barb Fitting, Cross Union, Polypropylene, 1/4"; 25/Pk EW-50143-23 Mfr # PFX250PPAF
COMPARE
1/4
$277.75USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hobe Barb Fitting, Cross Union, Polypropylene, 3/8"; 25/Pk EW-50143-35 Mfr # PFX375PPAF
COMPARE
3/8
$292.75USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hobe Barb Fitting, Cross Union, Polypropylene, 1/2"; 25/Pk EW-50143-43 Mfr # PFX500PPAF
COMPARE
1/2
$307.50USD / Pkg of 25