Saint-Gobain Pure-Fit® SIB® Hose Barb Fittings, Straight Reducer, Polypropylene
Smooth inner bore ensures seamless fluid transition between tubing and fittings
  • Unique internal design eliminates dead space
  • No areas of entrapment—prohibiting a potential for bacterial growth
  • Use with Pure-Fit and BarbLock® retainers for leak-free connections
  • Easily connect two different size tubing with a reducing fitting
  • Steriliza by gamma irradiation
  • Animal-derived component free and BPA free
  • Manufactured and packaged in Class 8 cleanroom
  • USP <88> Class VI and USP <87> compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Eliminate dead space and entrapment, a potential breeding ground for bacterial growth, with smooth inner bore (SIB) fittings. SIB fittings provide a seamless, “zero” transition between tubing and fittings, maintaining complete fluid integrity. Use with Pure-Fit or BarbLock retainer assembly that provides 360° compression to ensure a leak-free connection.
$225.50 - $282.25USD / Pkg of 25
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in)
Availability Pricing
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 1/8" x 1/16"; 25/Pk EW-50143-07 Mfr # PFR062X125PPAF
COMPARE
1/8 1/16
$234.75USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 1/4" x 1/16" ; 25/Pk EW-50143-08 Mfr # PFR062X250PPAF
COMPARE
1/4 1/16
$225.50USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 3/16" x 1/8"; 25/Pk EW-50143-13 Mfr # PFR125X187PPAF
COMPARE
3/16 1/8
$247.25USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 1/4" x 1/8"; 25/Pk EW-50143-14 Mfr # PFR125X250PPAF
COMPARE
1/4 1/8
$237.50USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 5/16" x 3/16"; 25/Pk EW-50143-19 Mfr # PFR187X312PPAF
COMPARE
5/16 3/16
$254.25USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 3/8" x 3/16"; 25/Pk EW-50143-20 Mfr # PFR187X375PPAF
COMPARE
3/8 3/16
$258.00USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 3/8" x 1/4"; 25/Pk EW-50143-28 Mfr # PFR250X375PPAF
COMPARE
3/8 1/4
$255.50USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 1/2" x 3/8"; 25/Pk EW-50143-40 Mfr # PFR375X500PPAF
COMPARE
1/2 3/8
$268.50USD / Pkg of 25
Saint Gobain Pure-Fit® SIB® Hose Barb Fitting, Straight Reducer, Polypropylene, 1/2" x 1/4"; 25/Pk EW-50143-48 Mfr # PFR250X500PPAF
COMPARE
1/2 1/4
$282.25USD / Pkg of 25
Showing 1- 9 of 9