Salisbury by Honeywell Electrical Insulating Rubber Glove Kits
Comply with NFPA 70E standard while protecting employees from dangerous electrical injuries
Get everything you need to protect yourself and keep your insulating rubber gloves in top working condition. Kits include a pair of insulating rubber gloves; the appropriate leather protector that protects the glove from cuts, abrasions, and punctures; and a storage bag which keeps the glove flat, preventing folds and creases that strain the rubber, shortening the life of the glove.
Note: In accordance with NFPA 70E, anything over 50 volts requires insulating rubber gloves.
$181.50 - $446.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compliance Size
Availability Pricing
Honeywell GK0011R8 Salisbury Insulated Glove Kit, Class 00, Red, 11"L, Size 8; One Pair EW-86435-00 Mfr # GK0011R/8
COMPARE
Red Class 00 8
$210.75USD / Each
Honeywell GK0011R9 Salisbury Insulated Glove Kit, Class 00, Red, 11"L, Size 9; One Pair EW-86435-01 Mfr # GK0011R9
COMPARE
Red Class 00 9
$229.25USD / Each
Honeywell GK0011R11 Salisbury Insulated Glove Kit, Class 00, Red, 11"L, Size 11; One Pair EW-86435-03 Mfr # GK0011R/11
COMPARE
Red Class 00 11
$225.00USD / Each
Honeywell GK011R8 Salisbury Insulated Glove Kit, Class 0, Red, 11"L, Size 8; One Pair EW-86435-10 Mfr # GK011R/8
COMPARE
Red Class 0 8
$281.25USD / Each
Honeywell GK011R10 Salisbury Insulated Glove Kit, Class 0, Red, 11"L, Size 10; One Pair EW-86435-12 Mfr # GK011R10
COMPARE
Red Class 0 10
$229.50USD / Each
Honeywell GK011B11 Salisbury Insulated Glove Kit, Class 0, Black, 11"L, Size 11; One Pair EW-86435-18 Mfr # GK011B11
COMPARE
Black Class 0 11
$203.75USD / Each
Honeywell GK011B12 Salisbury Insulated Glove Kit, Class 0, Black, 11"L, Size 12; One Pair EW-86435-19 Mfr # GK011B12
COMPARE
Black Class 0 12
$181.50USD / Each
Honeywell GK011Y9 Salisbury Insulated Glove Kit, Class 0, Yellow, 11"L, Size 9; One Pair EW-86435-21 Mfr # GK011Y/9
COMPARE
Yellow Class 0 9
$233.50USD / Each
Honeywell GK011Y10 Salisbury Insulated Glove Kit, Class 0, Yellow, 11"L, Size 10; One Pair EW-86435-22 Mfr # GK011Y/10
COMPARE
Yellow Class 0 10
$233.50USD / Each
Honeywell GK011Y12 Salisbury Insulated Glove Kit, Class 0, Yellow, 11"L, Size 12; One Pair EW-86435-24 Mfr # GK011Y/12
COMPARE
Yellow Class 0 12
$233.50USD / Each
Honeywell GK214RB9 Salisbury Insulated Glove Kit, Class 2, Black/Red, 14"L, Size 9; One Pair EW-86435-26 Mfr # GK214RB/9
COMPARE
Black Class 2 9
$446.00USD / Each
Honeywell GK214RB11 Salisbury Insulated Glove Kit, Class 2, Black/Red, 14"L, Size 11; One Pair EW-86435-28 Mfr # GK214RB11
COMPARE
Black Class 2 11
$446.00USD / Each
Showing 1- 12 of 12