Sani-Tech® Ultra-C Platinum-Cured Sanitary Silicone Tubing
Ultra-pure silicone tubing with ultra-low extractable is perfect for biopharmaceutical applications
  • Offers excellent flexibility and superior pump life
  • Platinum cured silicone—shows exceptional biocompatibility
  • Low extractables—perfect for maintaining fluid purity
  • Meets the requirements of USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5
  • Laser etched part numbers and lot codes for traceability
  • Produced and double-bagged in an ISO Class 7 certified cleanroom
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection cleaning and sterilization
Manufactured from the finest grade of silicone material, Sani-Tech Ultra-C tubing offers the utmost purity in tubing. Extremely low TOCs ensure minimal contamination by extractables, making this perfect for biopharmaceutical application. Laser etched part numbers and lot codes provide product traceability.
$46.70 - $1,190.00USD / Pkg of 1
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/32" ID x 1/16" OD; 25 Ft EW-50202-33 Mfr # ULTRA-C-030-0
COMPARE
1/32 1/16 1/64
$48.80USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/16" ID x 1/8" OD; 25 Ft EW-50202-34 Mfr # ULTRA-C-062-1
COMPARE
1/16 1/8 1/32
$46.70USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/16" ID x 3/16" OD; 25 Ft EW-50202-35 Mfr # ULTRA-C-062-2
COMPARE
1/16 3/16 1/16
$62.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/16" ID x 5/16" OD; 25 Ft EW-50202-37 Mfr # ULTRA-C-062-4
COMPARE
1/16 5/16 1/8
$120.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1" ID x 1-1/4" OD; 25 Ft EW-50202-38 Mfr # ULTRA-C-1000-4
COMPARE
1 1 1/4 1/8
$405.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1" ID x 1-3/8" OD; 25 Ft EW-50202-39 Mfr # ULTRA-C-1000-5
COMPARE
1 1 3/8 3/16
$1,045.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1" ID x 1-1/2" OD; 25 Ft EW-50202-40 Mfr # ULTRA-C-1000-6
COMPARE
1 1 1/2 1/4
$1,190.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/8" ID x 3/16" OD; 25 Ft EW-50202-41 Mfr # ULTRA-C-125-1
COMPARE
1/8 3/16 1/32
$54.60USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/8" ID x 1/4" OD; 25 Ft EW-50202-42 Mfr # ULTRA-C-125-2
COMPARE
1/8 1/4 1/16
$71.40USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/8" ID x 5/16" OD; 25 Ft EW-50202-43 Mfr # ULTRA-C-125-3
COMPARE
1/8 5/16 3/32
$102.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/8" ID x 3/8" OD; 25 Ft EW-50202-44 Mfr # ULTRA-C-125-4
COMPARE
1/8 3/8 1/8
$125.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/8" ID x 1/2" OD; 25 Ft EW-50202-45 Mfr # ULTRA-C-125-5
COMPARE
1/8 1/2 3/16
$221.75USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 5/32" ID x 9/32" OD; 25 Ft EW-50202-47 Mfr # ULTRA-C-156-2
COMPARE
5/32 9/32 1/16
$82.60USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 5/32" ID x 13/32" OD; 25 Ft EW-50202-49 Mfr # ULTRA-C-156-4
COMPARE
5/32 13/32 1/8
$148.25USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 3/16" ID x 1/4" OD; 25 Ft EW-50202-50 Mfr # ULTRA-C-187-1
COMPARE
3/16 1/4 1/32
$54.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 3/16" ID x 5/16" OD; 25 Ft EW-50202-51 Mfr # ULTRA-C-187-2
COMPARE
3/16 5/16 1/16
$88.20USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 3/16" ID x 3/8" OD; 25 Ft EW-50202-52 Mfr # ULTRA-C-187-3
COMPARE
3/16 3/8 3/32
$120.75USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 3/16" ID x 7/16" OD; 25 Ft EW-50202-53 Mfr # ULTRA-C-187-4
COMPARE
3/16 7/16 1/8
$165.00USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/4" ID x 3/8" OD; 25 Ft EW-50202-55 Mfr # ULTRA-C-250-2
COMPARE
1/4 3/8 1/16
$103.25USD / Pkg of 1
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/4" ID x 7/16" OD; 25 Ft EW-50202-56 Mfr # ULTRA-C-250-3
COMPARE
1/4 7/16 3/32
$142.25USD / Pkg of 1
Showing 1 - 20 of 32