SaniSure® Cap2V8® Aseptic Centrifuge Tube Assemblies, Labcon® Tubes
Reduce contamination and product loss with one-piece molded tube and cap assemblies
  • Ideal for aseptic transfer, storage and sampling
  • Cost-effective—reduces assembly and installation time
  • Lot-traceable
  • Features two ports (vent and transfer) with 1/8"ID x 1/4"OD tubing
  • Meets USP Class VI requirements
The single-use centrifuge assemblies are ideal for aseptic storage, transfer, and sampling. One continuous flow path reduces contamination chances and product loss. Each assembly includes a centrifuge tube and molded cap with transfer and vent tubing.
$121.50 - $141.75USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features Sterile Tube Capacity (mL)
Availability Pricing
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly (Labcon®), TPE Tubing, 15 mL, Sterile EW-50142-67 Mfr # CAP2V8-15C-A-Y-GI
COMPARE
8"L transfer line and 2"L vent line (TPE tubing) Yes 15
$128.00USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly (Labcon®), TPE Tubing, 15 mL, Nonsterile EW-50142-68 Mfr # CAP2V8-15C-A-Y-NG
COMPARE
8"L transfer line and 2"L vent line (TPE tubing) No 15
$122.50USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly (Labcon®), TPE Tubing, 50 mL, Sterile EW-50142-69 Mfr # CAP2V8-50C-A-Y-GI
COMPARE
12"L transfer line and 4"L vent line (TPE tubing) Yes 50
$138.75USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly with Dip Tube (Labcon®), TPE Tubing, 50 mL, Sterile EW-50142-70 Mfr # CAP2V8-50C-A-Y-GIDI
COMPARE
Dip tube, 12"L transfer line, and 4"L vent line (TPE tubing) Yes 50
$141.75USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly (Labcon®), TPE Tubing, 50 mL, Nonsterile EW-50142-71 Mfr # CAP2V8-50C-A-Y-NG
COMPARE
12"L transfer line and 4"L vent line (TPE tubing) No 50
$121.50USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly with Dip Tube (Labcon®), TPE Tubing, 50 mL, Nonsterile EW-50142-72 Mfr # CAP2V8 -50C - A - Y -NG - DIP
COMPARE
Dip tube, 12"L transfer line, and 4"L vent line (TPE tubing) No 50
$123.50USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly (Labcon®), PVC Tubing, 50 mL, Sterile EW-50142-73 Mfr # CAP2V8 -50C - A - C -GI -LC
COMPARE
12"L transfer line and 4"L vent line (PVC tubing) Yes 50
$138.75USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly with Dip Tube (Labcon®), PVC Tubing, 50 mL, Sterile EW-50142-74 Mfr # CAP2V8 -50C - A - C -GI - DIP -LC
COMPARE
Dip tube, 12"L transfer line, and 4"L vent line (PVC tubing) Yes 50
$141.75USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly (Labcon®), PVC Tubing, 50 mL, Nonsterile EW-50142-75 Mfr # CAP2V8 -50C - A - C -NG - LC
COMPARE
12"L transfer line and 4"L vent line (PVC tubing) No 50
$121.50USD / Each
SaniSure Cap2V8® Aseptic Centrifuge Tube Assembly with Dip Tube (Labcon®), PVC Tubing, 50 mL, Nonsterile EW-50142-76 Mfr # CAP2V8 -50C - A - C -NG - DIP -LC
COMPARE
Dip tube, 12"L transfer line, and 4"L vent line (PVC tubing) No 50
$122.75USD / Each
Showing 1- 10 of 10