Sartorius Arium® Mini Essential Ultrapure Water Systems
Produces up to 10 L/day of ASTM Type I ultrapure water
  • Large touch display with self-explanatory navigation
  • Take up a small footprint—only measures 28 cm in width
  • Volume-controlled dispensing in 50 mL steps
  • Delivers up to 1 L/minute
Using high-quality water ensures the reproducibility of your results and prevents time-consuming repeat analysis. The Arium® Mini Essential ultrapure water systems connect directly to a pretreated DI, RO, or EDI feed water line to produce up to 10 L/day of ASTM Type I ultrapure (18.2 MO) water.
These compact Arium Mini systems only measure 28 cm (11") wide to easily fit on a benchtop. A high-resolution, touch-activated color display featuring self-explanatory icons is easy to navigate even while wearing gloves. Store favorite functions such repeated dispensing of identical volumes to help increase efficiency while minimizing errors.
Ultrapure water is excellent for general laboratory applications. Select Arium Mini Essential with UV lamp for analytical applications.
$6,098.24 - $6,927.78USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Power (VAC) With UV Lamp
Availability Pricing
Sartorius Arium® Mini Essential Benchtop Water Purification System EW-99293-70 Mfr # H2O-MU-T-US
COMPARE
Arium® Mini Essential System 100 to 240 No
$6,098.24USD / Each
Sartorius Arium® Mini Essential Benchtop Water Purification System with UV Lamp EW-99293-71 Mfr # H2O-MU-UV-T-US
COMPARE
Arium® Mini Essential System with UV Lamp 100 to 240 Yes
$6,927.78USD / Each