Sartorius Arium® Mini Series Starter Sets
Starter set includes all the required components to get you up and running
  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Large touch display with self-explanatory navigation
  • Take up a small footprint—only measures 28 cm in width
  • Volume-controlled dispensing in 50 mL steps
  • Delivers up to 1 L/minute
Using high-quality water ensures the reproducibility of your results and prevents time-consuming repeat analysis. These compact Arium Mini systems only measure 28 cm (11") wide to easily fit on a benchtop. A high-resolution, touch-activated color display featuring self-explanatory icons is easy to navigate even while wearing gloves. Store favorite functions such repeated dispensing of identical volumes to help increase efficiency while minimizing errors.
Select from Arium Mini, Mini Plus, and Mini Essential depending on your application. All models produce up to 10 L of ASTM Type I ultrapure water per day. Arium Mini and Mini Plus use a closed Bagtank system while Mini Essential has no storage system. For analytical applications, order the Arium Mini with UV lamp to reduce total organic compound (TOC) to = 5 ppb.
$5,598.27 - $9,093.55USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ASTM Water Grade Source With UV Lamp
Availability Pricing
Sartorius Arium® Mini Plus Starter Set EW-99294-44 Mfr # H2O-MA-SET-US
COMPARE
Type I & Type III Direct conection to portable water supply No
$8,219.17USD / Each
Sartorius Arium® Mini Plus Starter Set with UV Lamp EW-99294-45 Mfr # H2O-MA-UV-SET-US
COMPARE
Type I & Type III Direct conection to portable water supply Yes
$9,093.55USD / Each
Sartorius Arium® Mini Starter Set EW-99294-46 Mfr # H2O-MM-SET-US
COMPARE
Type I Manual feeed with pretreated water No
$5,598.27USD / Each
Sartorius Arium® Mini Starter Set with UV Lamp EW-99294-47 Mfr # H2O-MM-UV-SET-US
COMPARE
Type I Manual feeed with pretreated water Yes
$6,338.13USD / Each
Sartorius Arium® Mini Essential Starter Set EW-99294-48 Mfr # H2O-MU-SET-US
COMPARE
Type I Pretreated RO, DI, or EDI water line No
$6,711.43USD / Each
Sartorius Arium® Mini Essential Starter Set with UV Lamp EW-99294-49 Mfr # H2O-MU-UV-SET-US
COMPARE
Type I Pretreated RO, DI, or EDI water line Yes
$7,540.97USD / Each