Sartorius Arium® Smart Stations
Remotely dispense pure and ultrapure water from water purification system or Bagtank
  • Accurate volume dispensing for fast, precise buffer and sample preparation
  • Compact design easily fits into any lab environment
  • Easy-to-navigate touch-activated color display
  • Stepless height adjustment to fill different size containers
  • Adjust flow rate from drop-by-drop to 2 L/min
Combine the Arium smart station with a water purification system or a Bagtank to remotely dispense pure or ultrapure water. These compact smart stations take up minimal space for easy and space-saving integration in your lab work area.
Use the large touch-activated color display to access all dispense options and view important quality parameters. Flexible, stepless height adjustment and a full 360° rotating dispenser arm make it easy to dispense water into a broad range of containers. The ergonomic design of the dispenser arm supports left- and right-hand operation. Adjust dispense volume from drop-by-drop to 2 L/min to meet your buffer and sample preparation needs.
$2,556.25 - $3,253.40USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Compatible With Description
Availability Pricing
Sartorius Arium® Smart Station Pure Water Remote Dispenser, Wall Mount EW-99293-90 Mfr # H2O-ARST-P-B
COMPARE
100 to 240 Arium® Bagtanks Wall-Mount Pure Water Remote Dispenser
$2,556.25USD / Each
Sartorius Arium® Smart Station Pure Water Remote Dispenser, Benchtop EW-99293-91 Mfr # H2O-ARST-P-T
COMPARE
100 to 240 Arium® Bagtanks Benchtop Pure Water Remote Dispenser
$2,556.25USD / Each
Sartorius Arium® Smart Station Ultrapure Water Remote Dispenser, Wall Mount EW-99293-92 Mfr # H2O-ARST-UP-B
COMPARE
100 to 240 Arium® Pro and Arium Comfort Wall-Mount Ultrapure Water Remote Dispenser
$3,253.40USD / Each
Sartorius Arium® Smart Station Ultrapure Water Remote Dispenser, Benchtop EW-99293-93 Mfr # H2O-ARST-UP-T
COMPARE
100 to 240 Arium® Pro and Arium Comfort Benchtop Ultrapure Water Remote Dispenser
$3,253.40USD / Each