Sartorius® Cubis® II (MCE) High-Capacity Micro Analytical Balances
Offers error-free operation with outstanding accuracy and repeatability
  • Ultra-high resolution with up to 61 million weighing steps
  • End-to-end data integrity
  • 21 CFR Part 11 and EU Annex 11 compliance
  • Integrated audit-trail
  • Motorized auto-leveling
  • State-of-the-art user management
  • Innovative TFT touch screen operation
  • Tool-free assembly/disassembly for easy cleaning
  • High chemical resistance to most cleaning agents
These high-capacity balances feature an easy-to-read, large, high-contrast TFT touch screen, perfect for routine weighing tasks. The balances are simple to operate and allow you to weigh very small samples directly into large flasks with the highest level of accuracy and repeatability, making them ideal for use in pharmaceutical, biopharmaceutical, chemical, environmental, automotive, and academia industries.
The balances are less prone to environmental effects, resulting in faster weighing workflows and improved performance in a daily laboratory environment. A built-in intelligent compensating system manages temperature, humidity, and air pressure changes which further ensures stable balance behavior. A spacious weighing area supports ergonomic weighing into various flasks. Select models with a built-in ionizer for samples prone to static charges and automated draft shield for hands-free convenience.
ITEM(S) ON SALE
$32,798.62 - $48,205.34USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Repeatability (mg) Description
Availability Pricing
Sartorius MCE36S-3S00-D HWL Cubis® II High-Capacity Micro Balance, 32g x 0.001mg; Activated Ionizer, Motorized Draft Shield EW-39743-25 Mfr # MCE36S-3S00-D HWL
COMPARE
32 0.0015 High-Capacity Micro Balance, 32g x 0.001mg; Activated Ionizer, Motorized Draft Shield
$39,161.74USD / Each
$44,501.98USD / Each
Sartorius MCE36S-3S00-D Cubis® II High-Capacity Micro Balance, 32g x 0.001mg; Manual Draft Shield EW-39743-26 Mfr # MCE36S-3S00-D
COMPARE
32 0.0015 High-Capacity Micro Balance, 32g x 0.001mg; Manual Draft Shield
$38,255.46USD / Each
$43,472.12USD / Each
Sartorius MCE36P-3S00-D HWL Cubis® II High-Capacity Micro Balance, 10.1g/32g x 0.001mg/0.01mg; Activated Ionizer, Motorized Draft Shield EW-39743-27 Mfr # MCE36P-3S00-D HWL
COMPARE
32 0.002 High-Capacity Micro Balance, 10.1g/32g x 0.001mg/0.01mg; Activated Ionizer, Motorized Draft Shield
$33,704.89USD / Each
$38,301.02USD / Each
Sartorius MCE36P-3S00-D Cubis® II High-Capacity Micro Balance, 10.1g/32g x 0.001mg/0.01mg; Manual Draft Shield EW-39743-28 Mfr # MCE36P-3S00-D
COMPARE
32 0.002 High-Capacity Micro Balance, 10.1g/32g x 0.001mg/0.01mg; Manual Draft Shield
$32,798.62USD / Each
$37,271.16USD / Each
Sartorius MCE66S-3S00-D HWL Cubis® II High-Capacity Micro Balance, 61g x 0.001mg; Activated Ionizer, Motorized Draft Shield EW-39743-29 Mfr # MCE66S-3S00-D HWL
COMPARE
61 0.0015 High-Capacity Micro Balance, 61g x 0.001mg; Activated Ionizer, Motorized Draft Shield
$48,205.34USD / Each
$54,778.80USD / Each
Sartorius MCE66S-3S00-D Cubis® II High-Capacity Micro Balance, 61g x 0.001mg; Manual Draft Shield EW-39743-30 Mfr # MCE66S-3S00-D
COMPARE
61 0.0015 High-Capacity Micro Balance, 61g x 0.001mg; Manual Draft Shield
$47,299.06USD / Each
$53,748.94USD / Each
Sartorius MCE66P-3S00-D HWL Cubis® II High-Capacity Micro Balance, 12g/61g x 0.001mg/0.01mg; Activated Ionizer, Motorized Draft Shield EW-39743-31 Mfr # MCE66P-3S00-D HWL
COMPARE
61 0.002 High-Capacity Micro Balance, 12g/61g x 0.001mg/0.01mg; Activated Ionizer, Motorized Draft Shield
$44,091.13USD / Each
$50,103.56USD / Each
Sartorius MCE66P-3S00-D Cubis® II High-Capacity Micro Balance, 12g/61g x 0.001mg/0.01mg; Manual Draft Shield EW-39743-32 Mfr # MCE66P-3S00-D
COMPARE
61 0.002 High-Capacity Micro Balance, 12g/61g x 0.001mg/0.01mg; Manual Draft Shield
$43,184.84USD / Each
$49,073.69USD / Each
Showing 1- 8 of 8