Sartorius Entris® II NTEP-Approved Toploading Balances
High accuracy and fast stabilization time ensure excellent weighing result every time
  • NTEP-approved legal-for-trade balances offer quality precision weighing
  • LED touch technology and 12 built-in applications
  • Constructed of highly chemical-resistant materials
Sartorius Entris® II toploading balances offer the best value in its class. Constructed of durable polybutylene terephthalate (PBT), stainless steel, and glass, these balances provide excellent chemical resistance and are easy to clean. Quick wipe down design and easy-to-remove parts help prevent cross contamination.
Built-in applications such as dosing, counting, percentage weighing, mixing, totalizing, animal weighing, free factor, density determination, statistics, peakhold, checkweighing, mass unit conversion, and underfloor weighing help make your weighing process more efficient. Easily connect to your PC using the RS-232 or USB-C interfaces. They are “PC-direct”, meaning no extra software is required and you can download your data directly to Microsoft® Word or Excel.
ITEM(S) ON SALE
$1,246.73 - $2,608.54USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg)
Availability Pricing
Sartorius BCE323-1NUS Entris® II NTEP-Approved Toploading Balance, 320 g x 1 mg EW-55000-29 Mfr # BCE323-1NUS
COMPARE
320 1
$1,611.15USD / Each
$1,830.86USD / Each
Sartorius BCE623-1NUS Entris® II NTEP-Approved Toploading Balance, 620 g x 1 mg EW-55000-30 Mfr # BCE623-1NUS
COMPARE
620 1
$2,359.20USD / Each
$2,680.91USD / Each
Sartorius BCE653-1NUS Entris® II NTEP-Approved Toploading Balance, 650 g x 1 mg EW-55000-31 Mfr # BCE653-1NUS
COMPARE
650 1
$2,608.54USD / Each
$2,964.26USD / Each
Sartorius BCE622-1NUS Entris® II NTEP-Approved Toploading Balance, 620 g x 10 mg EW-55000-32 Mfr # BCE622-1NUS
COMPARE
620 10
$1,246.73USD / Each
$1,416.74USD / Each
Sartorius BCE1202-1NUS Entris® II NTEP-Approved Toploading Balance, 1,200 g x 10 mg EW-55000-33 Mfr # BCE1202-1NUS
COMPARE
1200 10
$1,496.07USD / Each
$1,700.09USD / Each
Sartorius BCE3202-1NUS Entris® II NTEP-Approved Toploading Balance, 3,200 g x 10 mg EW-55000-34 Mfr # BCE3202-1NUS
COMPARE
3200 10
$1,745.42USD / Each
$1,983.44USD / Each
Sartorius BCE4202-1NUS Entris® II NTEP-Approved Toploading Balance, 4,200 g x 10 mg EW-55000-35 Mfr # BCE4202-1NUS
COMPARE
4200 10
$1,985.18USD / Each
$2,255.89USD / Each
Sartorius BCE5201-1NUS Entris® II NTEP-Approved Toploading Balance, 5,200 g x 100 mg EW-55000-36 Mfr # BCE5201-1NUS
COMPARE
5200 100
$1,467.30USD / Each
$1,667.39USD / Each
Sartorius BCE6202-1NUS Entris® II NTEP-Approved Toploading Balance, 6,200 g x 10 mg EW-55000-37 Mfr # BCE6202-1NUS
COMPARE
6200 10
$2,608.54USD / Each
$2,964.26USD / Each
Sartorius BCE8201-1NUS Entris® II NTEP-Approved Toploading Balance, 8,200 g x 100 mg EW-55000-38 Mfr # BCE8201-1NUS
COMPARE
8200 100
$1,707.05USD / Each
$1,939.84USD / Each
Showing 1- 10 of 10