SAS Super 100 Digital Microbial Air Samplers
Accurately collect and quantify airborne bacteria, viruses, molds, and fungi
  • Choose from eight preset sample volumes (10, 20, 30, 50, 100, 200, 500, or 1000 liters)
  • Program up to eight custom sample volumes
  • Detachable metal cover with intake holes creates laminar air flow
  • Flow rate is 100 liters/minute
  • Set parameters and view readings on the easy-to-read digital display
  • Unit retains previous sample volume
  • Microbes are driven onto the surface of a contact plate for incubation and analysis (order contact plate 14006-50 separately below)
  • 180l/min microbial air samplers are available, contact our application specialists for details
Impaction-style SAS air samplers let you accurately collect and quantify airborne bacteria, viruses, molds, and fungi. Ideal for clean rooms and process areas in pharmaceutical, electronics, and food tech environments. Microbes are driven onto the surface of a contact plate for incubation and analysis. Able to sample 45,000 liters of air per battery charge. 180l/min microbial air samplers are available, contact our application specialists for details.
$11,585.00 - $11,935.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Power (VAC) Description
Availability Pricing
SAS 18198-00 Microbial Air Sampler, Digital, Stainless Steel Head; 110-220 VAC EW-33687-02 Mfr # 18198-20
COMPARE
Stainless steel head 115 Microbial air sampler, digital, stainless steel head, 110-220 VAC
$11,585.00USD / Each
SAS 18198-10 Microbial Air Sampler, Digital, Aluminum Head: 110-220 VAC EW-33687-12 Mfr # 18198-40
COMPARE
Aluminum head 115 Microbial air sampler, digital, aluminum head, 110-220 VAC
$11,935.00USD / Each