Scienceware CultureTemp 37°C Warming Plate
Maintain the ideal cell temperature outside the incubator
  • Optimal 37°C (±1°C) temperature reduces cell stress
  • Saves time by eliminating need to go back and forth from incubator to work area
Warming plate maintains a constant 37°C protecting cell viability and allowing trypsin to work efficiently without the need to return to the incubator repeatedly. Anodized aluminum top plate fits most tissue culture vessels and maintains a uniform temperature across the entire surface. The UV resistant polypropylene construction prevents the housing from breaking down under UV lights, is water resistant, and cleans easily with ethanol or IPA. A 15-minute countdown timer helps keep track of time sensitive applications and the small footprint makes it easy to use on the benchtop or in a cell culture hood.
$512.00 - $669.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scienceware F37015-0000 CultureTemp 37°C Warming Plate, 120 VAC EW-86578-15 Mfr # F37015-0000
COMPARE
120
$669.00USD / Each
Scienceware F37015-1000 CultureTemp 37°C Warming Plate, 220 VAC EW-86578-16 Mfr # F37015-1000
COMPARE
220
$512.00USD / Each