Scienceware Lab Companion Round Vacuum Desiccators
Maintain 29 in. Hg vacuum for over 72 hours at room temperature
  • Viton® three-way valve offers consistent and uniform vacuum draw, release, and gas exchange
  • Analog vacuum gauge allows quick and convenient checking of the vacuum level
  • Use with any vacuum with free air capcity of 1.5 cubic feet per minute (CFM) or greater
These desiccators provide premium vacuum sustainment for reliable sample storage or running experiments over extended periods. The transparent polycarbonate construction is durable, easy to clean, chemical resistant, and provides quick visualization of the contents. The amber tinted models block 100% of UVA, B, and C light, minimizing discoloration or damage to light-sensitive samples.
Traditional Round Desiccators feature a silicone O-ring that provides leakproof vacuum capability. A five-tab locking ring secures the top and bottom when the vacuum is not in use and provides extra security when moving the unit. Use the built-in vacuum gauge on the top of the unit to check the vacuum levels.
Six models to choose from; 6, 10, and 20 liter in clear or UV blocking amber.
$393.00 - $594.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chamber Volume (cu ft) Color Plate Size (mm)
Availability Pricing
Scienceware F42400-2021 Round Vacuum Desiccator, 6L. EW-08909-05 Mfr # F42400-2021
COMPARE
0.2 Clear 190
$439.00USD / Each
Scienceware F42400-2041 UV Blocking Round Vacuum Desiccator, 6L. EW-08909-07 Mfr # F42400-2041
COMPARE
0.2 Amber 190
$430.50USD / Each
Scienceware F42400-2121 Round Vacuum Desiccator, 10L. EW-08909-09 Mfr # F42400-2121
COMPARE
0.4 Clear 250
$393.00USD / Each
Scienceware F42400-2141 UV Blocking Round Vacuum Desiccator, 10L. EW-08909-11 Mfr # F42400-2141
COMPARE
0.4 Amber 250
$502.00USD / Each
Scienceware F42400-2221 Round Vacuum Desiccator, 20L. EW-08909-13 Mfr # F42400-2221
COMPARE
0.7 Clear 310
$483.00USD / Each
Scienceware F42400-2241 UV Blocking Round Vacuum Desiccator, 20L. EW-08909-15 Mfr # F42400-2241
COMPARE
0.7 Amber 310
$594.00USD / Each