Scroll Labs SVF Vacuum Pumps
Seal free, maintenance free, and oil free scroll vacuum pumps are innovative and easy to use
  • Their compact size is perfect for portable equipment
  • Can handle moisture and solvents without a ballast valve
  • Ultimate pressure can reach as low as 0.01 mbar
These scroll pumps feature patent protected floating scroll technology. Unlike standard scroll pumps that utilize a tip-seal to maintain sealing, these innovative scroll pumps are tip-seal free to eliminate downtime and expensive repairs.
Common applications that are perfect for these scroll pumps include rotary evaporation, distillation, botanical extraction, mass spectrometers, clean rooms, freeze drying, or gas analysis.
$3,390.00 - $4,300.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Max Vacuum (torr) Description
Availability Pricing
Scroll Labs SVF-E1-100C Scroll Pump, 100 L/min, < 0.1 mbar; US Plug, 100 to 240 VAC EW-79500-10 Mfr # SVF-E1-100C
COMPARE
3.5 0.075 US plug
$4,300.00USD / Each
Scroll Labs SVF-E2-100C Scroll Pump, 100 L/min, < 0.03 mbar; US Plug, 100 to 240 VAC EW-79500-12 Mfr # SVF-E2-100C
COMPARE
3.5 0.007 US plug
$4,290.00USD / Each
Scroll Labs SVF-E1-20C Scroll Pump, 20 L/min, < 0.1 mbar; US Plug, 100 to 240 VAC EW-79500-16 Mfr # SVF-E1-20C
COMPARE
0.7 0.075 US plug
$3,390.00USD / Each