Sellstrom Laboratory Faceshields
Ultimate in lightweight face protection
White ABS plastic crown includes ratchet headgear and deluxe perspiration band to maintain a cool, comfortable fit. Lightweight clear acetate window (included) is fog-free to allow maximum visibility. Ideal for laboratory use. Select single-crown faceshield for protection from splash and particles. Select double-crown faceshield with chin protector for added impact protection.
$109.25USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sellstrom 31140 Laboratory faceshield, double crown w/acetate shield
Buy More Save More
EW-86478-22 Mfr # S31140
COMPARE
Laboratory faceshield, double crown w/acetate shield
$109.25USD / Each