Setra Nema 4 -IP65 Gauge Pressure Transducer
  • FEATURES
  • High accuracy
  • NEMA 4/IP65
  • Wide operating temperature range
  • Compatible with a wide range of gases or liquids
  • Corrosive resistant all stainless steel wetted parts
  • Choice of voltage or current output
  • Operates on low cost unregulated power supply
  • Meets CE conformance standards
The Transducer is packaged in a die-cast aluminum enclosure that is specifically designed for NEMA4/IP65 service. The robust capacitive design is resistant to environmental effects, such as shock, vibration, temperature and EMI/RFI. Its stainless steel sensing element does not require isolation from corrosive media.
Available in a wide variety of gauge pressure ranges, it also features adjustable potentiometers for zero and span settings..Only 3.6” high x 4.0” wide, the unit is designed for compact installations. The removable cover provides easy access to the internal terminal strip for wiring. Installation is quick and easy with 1/2 inch internal threaded conduit ports for electrical termination.
ITEM(S) ON SALE
$545.70USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Min Pressure (PSI) Output
Availability Pricing
Setra 2561025PG2M11 NEMA 4/IP65 Pressure Transmitter, 25 psig, 4-20mA EW-68444-89 Mfr # 2561025PG2M11
COMPARE
25 0 4 to 20 mA (2-wire)
$545.70USD / Each
$642.00USD / Each
Setra 2561050PG2M11 NEMA 4/IP65 Pressure Transmitter, 50 psig, 4-20mA EW-68444-90 Mfr # 2561050PG2M11
COMPARE
50 0 4 to 20 mA (2-wire)
$545.70USD / Each
$642.00USD / Each
Setra 2561100PG2M11 100 Psig 4-20MA Nema4 Xmtr EW-68444-91 Mfr # 2561100PG2M11
COMPARE
100 0 4 to 20 mA (2-wire)
$545.70USD / Each
$642.00USD / Each
Setra 2561250PG2M11 250 Psig 4-20MA Nema4 Xmtr EW-68444-92 Mfr # 2561250PG2M11
COMPARE
250 0 4 to 20 mA (2-wire)
$545.70USD / Each
$642.00USD / Each
Setra 2561500PG2M11 500 Psig 4-20MA Nema4 Xmtr EW-68444-93 Mfr # 2561500PG2M11
COMPARE
500 0 4 to 20 mA (2-wire)
$545.70USD / Each
$642.00USD / Each