Shimpo MFD-Series Dual-Scale Mechanical Force Gauges
Dual scale (lb/kg) provides additional flexibility for multiple standard testing requirements
  • Mechanical design provides immediate response for instant analysis
  • Compact, lightweight, ergonomic design is perfect for handheld use
  • Easily mount on test stands with rear threaded inserts
These easy-to-use mechanical force gauges are all you need for your tension and compression force measurement applications. The gauges are perfect for both push and pull force testing in material testing, manufacturing, QC/QA, maintenance and inspection applications, and more.
Peak force operation and push-button reset allow you to quickly and easily conduct repetitive testing in an assembly line or in quality control applications. Both the real-time and peak function modes permit flexibility in viewing live readings or maximum recorded values.
The large dual scale displays readings in both pounds (lb) and kilograms (kg) reducing the chance of parallax reading errors and providing immediate measurement response times. Ergonomically designed, these gauges are extremely comfortable for handheld use, but you can also order the optional handle for loads over fifteen pounds.
$215.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Resolution (lbs)
Availability Pricing
Shimpo MFD-03 Mechanical Force Gauge, 11 lb EW-25301-72 Mfr # MFD-03
COMPARE
11 0.05
$215.00USD / Each
Shimpo MFD-05 Mechanical Force Gauge, 45 lb EW-25301-74 Mfr # MFD-05
COMPARE
45 0.2
$215.00USD / Each
Shimpo MFD-06 Mechanical Force Gauge, 110 lb EW-25301-75 Mfr # MFD-06
COMPARE
110 0.5
$215.00USD / Each