Sludge Judge 2 Sludge Samplers
Efficiently Measure settled solids in sewage treatment, chemical, and food production facilities
  • Determine settable solids levels up to 5% with a safe, non-caustic process
  • Complete three 5-ft (1.5-m) sections that screw together, or separate component pieces for custom lengths
Sludge Judge samplers come in different configurations to match your requirements. Choose a full assembly with three 5-ft (1.5-m) sections that screw together or customize the length with individual pieces. The bottom section has a ball valve that collects liquid upon submersion and seals when lifted. The middle section provides extension capabilities, and additional sections can be added. The top section includes a nylon rope for convenience. Each segment is marked in 1-ft (0.3-m) increments for accurate sludge volume measurement. The Sludge Judge II features a 1-1/4” (3.2 cm) diameter, doubling the sample capacity.
ITEM(S) ON SALE
$81.51 - $293.88USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sludge Judge Sampler System II, 1.25" OD, 15 ft
Always in Stock
EW-05465-30 Mfr # B01355
COMPARE
Sampler System II, 1.25" OD, 15 ft
$293.88USD / Each
$345.75USD / Each
Sludge Judge II C09250WA Replacement Bottom Section with Valve EW-05465-31 Mfr # B01356
COMPARE
replacement bottom section with valve
$142.16USD / Each
$167.25USD / Each
Sludge Judge B01357WA Replacement Middle/Extension Section
Always in Stock
EW-05465-32 Mfr # B01357
COMPARE
Replacement Middle/Extension Section
$113.81USD / Each
$133.90USD / Each
Sludge Judge Ultra C09252WA Replacement Top Section with Rope EW-05465-33 Mfr # B01358
COMPARE
Replacement Top Section with Rope
$81.51USD / Each
$95.90USD / Each