Sludge Judge Ultra Sludge Samplers
Efficiently Measure settled solids in sewage treatment, chemical, and food production facilities
  • Determine settable solids levels up to 5% with a safe, non-caustic process
  • Complete three 5-ft (1.5-m) sections that screw together, or separate component pieces for custom lengths
Sludge Judge samplers come in different configurations to match your requirements. Choose a full assembly with three 5-ft (1.5-m) sections that screw together or customize the length with individual pieces. The bottom section has a ball valve that collects liquid upon submersion and seals when lifted. The middle section provides extension capabilities, and additional sections can be added. The top section includes a nylon rope for convenience. Each segment is marked in 1-ft (0.3-m) increments for accurate sludge volume measurement. The Basic Sludge Judge and Sludge Judge Ultra have a 3/4" (1.9 cm) diameter and hold about 3 oz (89 mL) per foot. The Sludge Judge Ultra also includes UV stabilization to protect against sun damage.
ITEM(S) ON SALE
$96.85 - $260.86USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sludge Judge B01386WA Sampler System, 3/4" OD, 15 ft, with UV stabilizer
Always in Stock
EW-05465-40 Mfr # B01386
COMPARE
Sampler System, 3/4" OD, 15 ft, with UV stabilizer
$260.86USD / Each
$306.90USD / Each
Sludge Judge Ultra C09250WA Replacement Bottom Section with Valve EW-05465-41 Mfr # B01389
COMPARE
Replacement Bottom Section with Valve
$156.44USD / Each
$184.05USD / Each
Sludge Judge Ultra C09251WA Replacement Middle/Extension Section
Always in Stock
EW-05465-42 Mfr # B01388
COMPARE
Replacement Middle/Extension Section
$96.85USD / Each
$113.95USD / Each
Sludge Judge C09252WA Replacement Top Section with Rope EW-05465-43 Mfr # B01387
COMPARE
Replacement Top Section with Rope
$97.75USD / Each
$115.00USD / Each