Spex Assurance® Grade Aluminum, 1,000 µg/mL

Single-element standards for AA and ICP analysis
  • Made with acid and ASTM Type I water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. They are packaged in 30 mL, 125 mL, 250 mL, and 500 mL bottles to minimize contamination.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
$44.00 - $170.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Concentration Matrix Volume (mL)
Availability Pricing
COMPARE
1,000 µg/mL (1,000 ppm) 2% HNO3 125
$72.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) 2% HNO3 250
$126.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) 2% HNO3 30
$44.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) 2% HNO3 500
$163.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) 2% HCl 500
$170.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat