Spex Assurance® Grade Arsenic +3 Speciation Standards, 1,000 µg/mL
Speciation standards for analyzing species in a sample
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Speciation analysis has become common in many fields, including environmental, food and pharmaceutical testing labs. To analyze species within a sample requires Certified Reference Materials (CRMs) for sample verification and method validation. Many speciation standards are available in today’s market, however, most of them are not certified or analyzed with a state-of-the-art ICP, ICP-MS or LC-ICP-MS. SPEX CertiPrep offers a wide variety of speciation standards, certified to the strictest ISO 17034 guidelines, and tested on our own LC-ICP-MS.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$39.00 - $77.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
1,000 µg/mL (1,000 ppm) 125 Assurance® Grade Arsenic +3 Speciation Standard, 1,000 µg/mL in 2% HCl; 125 mL
$77.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) 30 Assurance® Grade Arsenic +3 Speciation Standard, 1,000 µg/mL in 2% HCl; 30 mL
$39.00USD / Each