Spex Assurance® Grade Secondary Drinking Water Metals for AA & ICP
Multi-element drinking water pollutant standards for AA & ICP analysis
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Refer to US National Primary Drinking Water Regulations 40 CFR, Part 141
Use these standards with procedures for compliance monitoring of drinking water and for analysis of ground and surface water where determination of the drinking water contamination levels are required.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$205.00 - $333.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Copper, Iron, Manganese, and Zinc 125 Spex CertiPrep Assurance® Grade Secondary Drinking Water Metals for AA & ICP in 2% HNO3; 125 mL
$205.00USD / Each
COMPARE
Copper, Iron, Manganese, and Zinc 500 Spex CertiPrep Assurance® Grade Secondary Drinking Water Metals for AA & ICP in 2% HNO3; 500 mL
$333.50USD / Each