Spex Assurance® Grade Silicon, 1,000 µg/mL
Single-element standards for AA and ICP analysis
  • Made with acid and ASTM Type I water
  • Inorganic compounds at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Custom standards can be manufactured upon request.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
$39.00 - $170.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Concentration Matrix Volume (mL)
Availability Pricing
COMPARE
1,000 µg/mL in water & 0.4% F Water & 0.4% fluoride 125
$76.00USD / Each
COMPARE
1,000 µg/mL in water & 0.4% F Water & 0.4% fluoride 250
$126.00USD / Each
COMPARE
1,000 µg/mL in water & 0.4% F Water & 0.4% fluoride 30
$39.00USD / Each
COMPARE
1,000 µg/mL in water & 0.4% F Water & 0.4% fluoride 500
$170.00USD / Each
COMPARE
1,000 µg/mL in water Water & 0.4% fluoride 500
$170.00USD / Each