Spex Assurance® Grade Trace Metals for AA & ICP in 5% HNO3
Multi-element standards for AA and ICP analysis
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • May be used either as standards or as a means to check the individual analysts accuracy and precision
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
Refer to US EPA Methods Manual 600/4-79-020 "Methods for Chemical Analysis of Water and Wastes" Trace Metals 21I, 21II and 21III Methods.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$307.00 - $617.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Assurance® Grade Trace Metals with Mercury for AA & ICP in 5% HNO3; 125 mL
$370.00USD / Each
COMPARE
Multiple 500 Assurance® Grade Trace Metals with Mercury for AA & ICP in 5% HNO3; 500 mL
$617.00USD / Each
COMPARE
Multiple 125 Assurance® Grade Trace Metals without Mercury for AA & ICP in 5% HNO3; 125 mL
$307.00USD / Each
COMPARE
Multiple 500 Assurance® Grade Trace Metals without Mercury for AA & ICP in 5% HNO3; 500 mL
$453.00USD / Each