Spex Assurance® Grade Trace Metals Standard Sets for AA & ICP
Multi-element groundwater and wastewater pollution control check standards for AA & ICP analysis
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Refer to US EPA Methods Manual 600/4-79-020 "Methods for Chemical Analysis of Water and Wastes" Trace Metals 21I, 21II and 21III Methods
May be used either as standards or as a means to check the individual analysts accuracy and precision.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$528.00 - $607.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Four bottle set 125 Spex CertiPrep Assurance® Grade Trace Metals Standard Set with Mercury for AA & ICP; 125 mL
$607.00USD / Each
COMPARE
Three bottle set 125 Spex CertiPrep Assurance® Grade Trace Metals Standard Set without Mercury for AA & ICP; 125mL
$528.00USD / Each