Spex Assurance® Grade Ytterbium, 1,000 µg/mL for AA and ICP
Single-element standards for AA and ICP analysis
  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$44.00 - $182.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Element/Ion Volume (mL)
Availability Pricing
COMPARE
1,000 µg/mL (1,000 ppm) Ytterbium (Yb) 125
$84.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) Ytterbium (Yb) 30
$44.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) Ytterbium (Yb) 500
$182.00USD / Each